Temperature Monitoring Systems, Ace Glass Incorporated
Supplier: Ace Glass
8318-115 8318-117 8318-119 8318-121 8318-123 8318-142 8318-144 8318-146 8318-148 8318-150 8318-20 8318-204 8318-207 8318-209 8318-35
89062-956EA 2241.98 CAD
89062-956 89062-958 89062-960 89062-962 89062-964 89062-966 89062-968 89062-970 89062-972 89062-974 89062-976 89062-978 89062-980 89062-982 89062-984
Temperature Monitoring Systems, Ace Glass Incorporated
Monitoring Systems CO2 and Temperature Monitoring Systems
This monitoring system is a complete package to enable the user to monitor the temperature of jacketed 10, 30, and 50 L cylindrical Pilot Plant Reactor
Complete with LED display and recorder output, PTFE covered Type J Temperature Sensor with detachable lead, PTFE Adapter, adapters to secure temperature sensor in head of reactor, and Nylon Bushing. Thermocouple only can be ordered.
Ordering information: The Complete Temperature Monitoring System for 10 or 20 mL Cylindrical comes with Thermometer, #7 Adapter, "J" Thermocouple Sensor, Sensor Cord. The Complete Temperature MonitoringMore Product Information