Spherical Joint Stopper Cap, Ace Glass Incorporated
Supplier: Ace Glass
8274-06 8274-96
89062-890EA 77.22 CAD
89062-890 89062-892
Spherical Joint Stopper Cap, Ace Glass Incorporated
Stoppers Glass Stoppers
Cap-type stoppers have spherical joints and are used to seal unused ports on Air Sampling Manifolds
Use pinch clamp to secure caps to ports.
Order Now
SPECIFICATIONS
Environmentally Preferable