Straight Bore Stopcock, Glass Plug, Ace Glass
Supplier: Ace Glass
8137-02 8137-04 8137-05 8137-06 8137-08 8137-09 8137-10 8137-12 8137-14
89062-312EA 172.23 CAD
89062-312 89062-314 89062-316 89062-318 89062-320 89062-322 89062-324 89062-326 89062-328
Straight Bore Stopcock, Glass Plug, Ace Glass
Stopcocks
A ruggedly constructed stopcock with solid glass plug and tapered flange which insures maximum strength at the seal
Bore of side arms is equal to or slightly larger than bores of plug on all sizes.
Order Now
SPECIFICATIONS
Environmentally Preferable