Beaded Process Pipe Short End Cap, Ace Glass Incorporated
Supplier: Ace Glass
8847-02 8847-04 8847-06 8847-08 8847-10
89064-634EA 100.37 CAD
89064-634 89064-636 89064-638 89064-640 89064-642
Beaded Process Pipe Short End Cap, Ace Glass Incorporated
Fittings and Connectors
This Beaded Process Pipe Short End Cap is made from Borosilicate glass.
  • The thermal expansion is 32 × 10-7/ °C
  • The shock difference 160 °C
In regards to durability acids, all except Hydrofluoric or Phosphoric with Fluorides. Some durability alkalies attack above 120 °F and above pH11.
Conductivity: 0.73 BTU/ hr. ft.2/F°/ft. at 212 °F