Methyl palmitate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

M-Clarity™ quality level = MQ200

8.43546.0100 8.43546.0500
CA8.43546.0100CS 1070 CAD
CA8.43546.0100 CA8.43546.0500
Methyl palmitate for synthesis, Sigma-Aldrich®
Methyl palmitate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 417
Melting Pt: 29.9
Density: 0.8804 g/cm³ (20 °C)
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 30 °C
Molar Mass 270.46 g/mol
Boiling Point 415 - 418 °C (996 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 113 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR