2,5-Dihydroxybenzoic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.41745.0050
8.41745.0250
CA8.41745.0050EA
103
CAD
CA8.41745.0050
CA8.41745.0250
2,5-Dihydroxybenzoic acid for synthesis, Sigma-Aldrich®
2,5-Dihydroxybenzoic acid
Formula:
C₇H₆O₄ Boiling Pt: 339 °C (1013 hPa) Melting Pt: 202…206 °C Density: 1.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002460 CAS Number: 490-79-9 EINECS: 207-718-5 Merck Index: 13,04409 |
Specification Test Results
Melting Point | 202 - 206 °C |
Molar Mass | 154.12 g/mol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...