Creatine (N-amidinosarcosine) monohydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate

M-Clarity™ quality level = MQ200

8.41470.0050 8.41470.0250
CA8.41470.0050EA 103 CAD
CA8.41470.0050 CA8.41470.0250
Creatine (N-amidinosarcosine) monohydrate for synthesis, Sigma-Aldrich®
Creatine (N-Amidinosarcosine) monohydrate
Formula: C₄H₉N₃O₂·1H₂O
MW: 149.15 g/mol (131.13 anhy)
MDL Number: MFCD00071582
CAS Number: 6020-87-7
EINECS: 200-306-6

Order Now

Specification Test Results

Solubility 17 g/l (20 °C)
Melting Point 296 - 297 °C (decomposition)
Molar Mass 149.15 g/mol
pH 6.9 (10 g/l, H2O, 20 °C)
Ignition Point 800 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR