DL-Camphor for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41456.0250 8.41456.1000
CA8.41456.0250CS 1335 CAD
CA8.41456.0250 CA8.41456.1000
DL-Camphor for synthesis, Sigma-Aldrich®
DL-Camphor
Formula: C₁₀H₁₆O
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III

Order Now

Specification Test Results

Solubility 1.5 g/l (20 °C)
Melting Point 180 °C
Molar Mass 152.24 g/mol
Bulk Density 800 kg/m3
Boiling Point 205 °C (1013 hPa)
Vapor Pressure 1.2 hPa (20 °C)
Flash Point 66 °C
Explosion Limit 0.6 - 4.5 %(V)
Density 0.99 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 460 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR