Span® 60 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.40121.0100 8.40121.0500
CA8.40121.0100EA 55 CAD
CA8.40121.0100 CA8.40121.0500
Span® 60 for synthesis, Sigma-Aldrich®
Span® 60
Formula: C₂₄H₄₆O₆
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS Number: 1338-41-6
EINECS: 215-664-9
Merck Index: 12,08872

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 53 °C
Molar Mass 430.62 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure  - 1.4 hPa (20 °C)
Flash Point 149 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR