2-Ethylhexyl methacrylate stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Methacrylic acid 2-ethylhexyl ester

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.40088.0025 8.40088.0250
CA8.40088.0025EA 59 CAD
CA8.40088.0025 CA8.40088.0250
2-Ethylhexyl methacrylate stabilised for synthesis, Sigma-Aldrich®
2-Ethylhexyl methacrylate
Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
MDL Number: MFCD00009494
CAS Number: 688-84-6
EINECS: 211-708-6

Order Now

Specification Test Results

Solubility  - 0.1 g/l (20 °C)
Melting Point -50 °C
Molar Mass 198.3 g/mol
Boiling Point 218 °C (1013 hPa)
Vapor Pressure 0.13 hPa (20 °C)
Flash Point 92 °C
Explosion Limit 0.6 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 380 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR