cis-Platin Pt 65% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride

M-Clarity™ quality level = MQ200

8.24510.0100 8.24510.0500
CA8.24510.0100EA 100 CAD
CA8.24510.0100 CA8.24510.0500
cis-Platin Pt 65% for synthesis, Sigma-Aldrich®
cis-Platin
Formula: H₆Cl₂N₂Pt
Density: 3.738 g/cm³ (20 °C)
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Merck Index: 13,02341

Order Now

Specification Test Results

Solubility 2.5 g/l (20 °C)
Molar Mass 300.05 g/mol
Bulk Density 850 kg/m3
Density 3.7 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR