2,4-Diaminophenol dihydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol

M-Clarity™ quality level = MQ200

8.22331.0100 8.22331.0250
CA8.22331.0100CS 1928 CAD
CA8.22331.0100 CA8.22331.0250
2,4-Diaminophenol dihydrochloride for synthesis, Sigma-Aldrich®
2,4-Diaminophenol dihydrochloride
Formula: C₆H₁₀Cl₂N₂O
MW: 197.06 g/mol
MDL Number: MFCD00012979
CAS Number: 137-09-7
EINECS: 205-279-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 275 g/l (15 °C)
Melting Point 205 °C
Molar Mass 197.06 g/mol
Bulk Density 310 kg/m3
pH 1 - 2 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR