1-Naphthylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.22291.0100 8.22291.1000
CA8.22291.0100CS 372 CAD
CA8.22291.0100 CA8.22291.1000
1-Naphthylamine for synthesis, Sigma-Aldrich®
1-Naphthylamine
Formula: C₁₀H₉N
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 50 °C
Density: 1.171 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00004016
CAS Number: 134-32-7
EINECS: 205-138-7
UN: 2077
ADR: 6.1,III
Merck Index: 12,06485

Order Now

Specification Test Results

Solubility 1.7 g/l (20 °C)
Melting Point 48 - 50 °C
Molar Mass 143.19 g/mol
Bulk Density 560 kg/m3
Boiling Point 7.1 (1 g/l, H2O, 20 °C)
Vapor Pressure 301 °C (1013 hPa)
Flash Point 0.003 hPa (20 °C)
Refractive Index 157 °C
Density 1.15 g/cm3 (20 °C)
Ignition Point 460 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR