Chromium hexacarbonyl for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Chromium carbonyl, Hexacarbonylchromium, Hexacarbonylchromium(0), Chromium(0) hexacarbonyl

M-Clarity™ quality level = MQ200

8.22196.0010 8.22196.0025
CA8.22196.0010EA 484 CAD
CA8.22196.0010 CA8.22196.0025
Chromium hexacarbonyl for synthesis, Sigma-Aldrich®
Chromium hexacarbonyl
Formula: C₆CrO₆
MW: 220.06 g/mol
Boiling Pt: 210
Melting Pt: 90
Density: 1.77 g/cm³ (20 °C)
MDL Number: MFCD00010945
CAS Number: 13007-92-6
EINECS: 235-852-4
UN: 3466
ADR: 6.1 III
Merck Index: 13,02253

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 149 - 150 °C
Molar Mass 220.06 g/mol
Boiling Point 151 °C (1013 hPa)
Vapor Pressure 0.35 hPa (20 °C)
Density 1.77 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR