1-Dodecene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.22048.0100 8.22048.0500
CA8.22048.0100CS 1295 CAD
CA8.22048.0100 CA8.22048.0500
1-Dodecene for synthesis, Sigma-Aldrich®
1-Dodecene
Formula: C₁₂H₂₄
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.7582 g/cm³ (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -35 °C
Molar Mass 168.32 g/mol
Boiling Point 213 - 216 °C (1013 hPa)
Vapor Pressure 0.23 hPa (20 °C)
Flash Point 77 °C
Refractive Index 1.4294 (20 °C, 589 nm)
Explosion Limit 0.6 - 5.4 %(V)
Density 0.76 g/cm3 (20 °C)
Ignition Point 226 °C
Viscosity kinematic 1.71 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR