4'-Bromoacetophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21968.0025 8.21968.0100
CA8.21968.0025EA 59 CAD
CA8.21968.0025 CA8.21968.0100
4'-Bromoacetophenone for synthesis, Sigma-Aldrich®
4'-Bromoacetophenone
Formula: C₈H₇BrO
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 49…52 °C
Density: 1.736 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000105
CAS Number: 99-90-1
EINECS: 202-799-3
UN: 2811
ADR: 6.1,II
Merck Index: 13,01387

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 49 - 52 °C
Molar Mass 199.04 g/mol
Boiling Point 255 °C (1013 hPa)
Flash Point 100 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR