Propyl benzoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21958.0005 8.21958.0250
CA8.21958.0005EA 41 CAD
CA8.21958.0005 CA8.21958.0250
Propyl benzoate for synthesis, Sigma-Aldrich®
Propyl benzoate
Formula: C₁₀H₁₂O₂
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00009370
CAS Number: 2315-68-6
EINECS: 219-020-8

Order Now

Specification Test Results

Solubility 0.35 g/l (25 °C)
Melting Point -52 °C
Molar Mass 164.2 g/mol
Boiling Point 231 °C (1013 hPa)
Vapor Pressure 0.181 hPa (25 °C)
Flash Point 100 °C
Refractive Index 1.4981 (25 °C, 589 nm)
Density 1.02 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR