2,3-Dimethylaniline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,3-Xylidine, 3-Amino-o-xylene

M-Clarity™ quality level = MQ200

8.21222.0005 8.21222.0250
CA8.21222.0005EA 41 CAD
CA8.21222.0005 CA8.21222.0250
2,3-Dimethylaniline for synthesis, Sigma-Aldrich®
2,3-Dimethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007732
CAS Number: 87-59-2
EINECS: 201-755-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Specification Test Results

Solubility 1.5 g/l (20 °C)
Melting Point 2 °C
Molar Mass 121.18 g/mol
Boiling Point 224 °C
Vapor Pressure 0.13 hPa (25 °C)
Flash Point 96 °C
Refractive Index 1.5685 (20 °C, 589 nm)
Explosion Limit 1 - 2.7 %(V)
Density 0.99 g/cm3 (20 °C)
Ignition Point 545 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR