Trimethylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Trimethylamine hydrochloride

M-Clarity™ quality level = MQ200

8.21178.0005 8.21178.0250
CA8.21178.0005EA 45 CAD
CA8.21178.0005 CA8.21178.0250
Trimethylammonium chloride for synthesis, Sigma-Aldrich®
Trimethylammonium chloride
Formula: C₃H₁₀ClN
MW: 95.57 g/mol
Melting Pt: 273…278 °C
Density: ∼1 g/cm³ (20 °C)
MDL Number: MFCD00012478
CAS Number: 593-81-7
EINECS: 209-810-0
Merck Index: 12,09841

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 273 - 278 °C
Molar Mass 95.57 g/mol
pH 5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR