Trimethylamine 40% in water for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.21177.0100 8.21177.1000 8.21177.2500
CA8.21177.0100CS 1003 CAD
CA8.21177.0100 CA8.21177.1000 CA8.21177.2500
Trimethylamine 40% in water for synthesis, Sigma-Aldrich®
Trimethylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 2.87
Melting Pt: -117.1
Density: 0.88 g/cm³ (20 °C)
Flash Pt: -20
Storage Temperature: Ambient
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 1297
ADR: 3 (8) I
Merck Index: 12,09841

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -2 °C
Boiling Point 30 - 100 °C
Vapor Pressure 600 hPa (20 °C)
Flash Point -45 °C
Explosion Limit 2 - 11.6 %(V) (anhydrous substance)
Density 0.88 g/cm3 (20 °C)
pH 13 (H2O, 20 °C)
Ignition Point 250 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR