Tetrahydrothiophene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: THT

M-Clarity™ quality level = MQ200

8.21095.0005 8.21095.0250
CA8.21095.0005EA 49 CAD
CA8.21095.0005 CA8.21095.0250
Tetrahydrothiophene for synthesis, Sigma-Aldrich®
Tetrahydrothiophene
Formula: C₄H₈S
MDL Number: MFCD00005476
CAS Number: 110-01-0
UN: 2412
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -96 °C
Molar Mass 88.16 g/mol
Boiling Point 121 °C (1013 hPa)
Vapor Pressure 19 hPa (20 °C)
Flash Point 19 °C
Refractive Index 1.504 (20 °C, 589 nm)
Explosion Limit 1.1 - 12.1 %(V)
Density 1.00 g/cm3 (20 °C)
pH 7 (H2O, 20 °C)
Ignition Point 202 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR