DL-Malic acid disodium salt for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Di-Sodium Dl-Malate, DL-Malic acid disodium salt, DL-Apple acid disodium salt, Disodium DL-Malate hydrate

M-Clarity™ quality level = MQ200

8.20872.0025 8.20872.0100
CA8.20872.0025EA 108 CAD
CA8.20872.0025 CA8.20872.0100
DL-Malic acid disodium salt for synthesis, Sigma-Aldrich®
DL-Malic acid disodium salt
Formula: C₄H₄Na₂O₅
MDL Number: MFCD00012466
CAS Number: 676-46-0

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 178.05 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR