1,1,3,3-Tetramethoxypropane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Malonaldehyde bis(dimethyl acetal)

M-Clarity™ quality level = MQ200

8.20756.0005 8.20756.0250
CA8.20756.0005EA 41 CAD
CA8.20756.0005 CA8.20756.0250
1,1,3,3-Tetramethoxypropane for synthesis, Sigma-Aldrich®
1,1,3,3-Tetramethoxypropane
Formula: C₇H₁₆O₄
MW: 164.2 g/mol
MDL Number: MFCD00008488
CAS Number: 102-52-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Molar Mass 164.2 g/mol
Boiling Point 183 °C
Vapor Pressure 1.7 hPa (20 °C)
Flash Point 66 °C
Refractive Index 1.4067 (20 °C, 589 nm)
Density 0.99 g/cm3 (20 °C)
pH 7 (H2O)
Viscosity kinematic 1.75 mm2/s (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR