Methyl hexanoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20638.0005 8.20638.0250
CA8.20638.0005EA 31 CAD
CA8.20638.0005 CA8.20638.0250
Methyl hexanoate for synthesis, Sigma-Aldrich®
Methyl hexanoate
Formula: C₇H₁₄O₂
MDL Number: MFCD00009510
CAS Number: 106-70-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Solubility 1.33 g/l (20 °C)
Melting Point -71 °C
Molar Mass 130.19 g/mol
Boiling Point 151 °C (1013 hPa)
Vapor Pressure 3.7 hPa (20 °C)
Flash Point 41 °C
Refractive Index 1.396 (20 °C, 589 nm)
Density 0.88 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR