Isobutyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Methylpropyl acetate

M-Clarity™ quality level = MQ200

8.20557.0100 8.20557.1000
CA8.20557.0100CS 538 CAD
CA8.20557.0100 CA8.20557.1000
Isobutyl acetate for synthesis, Sigma-Aldrich®
Isobutyl acetate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00008932
CAS Number: 110-19-0
UN: 1213
ADR: 3,II

Order Now

Specification Test Results

Solubility 7 g/l (20 °C)
Melting Point -99 °C
Molar Mass 116.16 g/mol
Boiling Point 116 - 118 °C (1013 hPa)
Vapor Pressure 17 hPa (20 °C)
Flash Point 18 °C
Refractive Index 1.3877 (20 °C, 589 nm)
Explosion Limit 2.4 - 10.5 %(V)
Density 0.87 g/cm3 (20 °C)
pH 5 (4 g/l, H2O, 20 °C)
Ignition Point 510 °C
Evaporation number 8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR