1,2-Epoxybutane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,2-Butylene oxide, 1,2-Butylene Oxide, 1,2-Epoxybutane, Ethylethylene Oxide, Ethyloxirane

M-Clarity™ quality level = MQ200

8.20551.0100 8.20551.1000 8.20551.2500
CA8.20551.0100CS 772 CAD
CA8.20551.0100 CA8.20551.1000 CA8.20551.2500
1,2-Epoxybutane for synthesis, Sigma-Aldrich®
1,2-Epoxybutane
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 63
Melting Pt: -130
Flash Pt: -26
MDL Number: MFCD00005153
CAS Number: 106-88-7
UN: 3022
ADR: 3 II

Order Now

Specification Test Results

Solubility 59 g/l (20 °C)
Melting Point -130 °C
Molar Mass 72.1 g/mol
Boiling Point 63 °C (1013 hPa)
Vapor Pressure 188 hPa (20 °C)
Flash Point -26 °C
Refractive Index 1.3835 (20 °C, 589 nm)
Explosion Limit 1.7 - 19 %(V)
Density 0.83 g/cm3 (20 °C)
pH 7 (50 g/l, H2O, 20 °C)
Ignition Point 370 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR