9,10-Diphenylanthracene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20529.0001 8.20529.0005
CA8.20529.0001EA 93 CAD
CA8.20529.0001 CA8.20529.0005
9,10-Diphenylanthracene for synthesis, Sigma-Aldrich®
9,10-Diphenylanthracene
Formula: C₂₆H₁₈
MDL Number: MFCD00001253
CAS Number: 1499-10-1

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 247 - 252 °C
Molar Mass 330.43 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR