Dibutyltin diacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20386.0100 8.20386.9010
CA8.20386.0100EA 145 CAD
CA8.20386.0100 CA8.20386.9010
Dibutyltin diacetate for synthesis, Sigma-Aldrich®
Dibutyltin diacetate
Formula: C₁₂H₂₄O₄Sn
MDL Number: MFCD00008697
CAS Number: 1067-33-0
UN: 2788
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 0.006 g/l (20 °C)
Melting Point 12 °C
Molar Mass 351 g/mol
Boiling Point 142 - 145 °C (13 hPa)
Vapor Pressure 1.7 hPa (25 °C)
Flash Point 143 °C
Density 1.30 g/cm3 (20 °C)
Ignition Point 200 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR