Tributyl citrate zS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Citric acid tributyl ester, Hydroxytricarballylic acid tributyl ester, Tributyl 2-hydroxypropane-1,2,3-tricarboxylate

MQ200

8.20350.0100 8.20350.1000
CA8.20350.0100CS 806 CAD
CA8.20350.0100 CA8.20350.1000
Tributyl citrate zS, Sigma-Aldrich®
Tributyl citrate
Formula: C₁₈H₃₂O₇
MW: 360.45 g/mol
Boiling Pt: 390
MDL Number: MFCD00027217
CAS Number: 77-94-1
Merck Index: 13,01563

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -20 °C
Molar Mass 360.44 g/mol
Boiling Point 169 - 171 °C (1 hPa)
Flash Point 184 °C
Refractive Index 1.443 (25 °C, 589 nm)
Density 1.04 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR