Sodium chloroacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20293.0100 8.20293.0500 8.20293.9050
CA8.20293.0100EA 58 CAD
CA8.20293.0100 CA8.20293.0500 CA8.20293.9050
Sodium chloroacetate for synthesis, Sigma-Aldrich®
Sodium chloroacetate
Formula: C₂H₂ClNaO₂
Storage Temperature: Ambient
MDL Number: MFCD00002684
CAS Number: 3926-62-3
UN: 2659
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 820 g/l (20 °C)
Molar Mass 116.47 g/mol
Bulk Density 850 kg/m3
Flash Point 270 °C
pH 4.5 - 9.0 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR