2-Chlorobutane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20281.0100 8.20281.1000
CA8.20281.0100CS 797 CAD
CA8.20281.0100 CA8.20281.1000
2-Chlorobutane for synthesis, Sigma-Aldrich®
2-Chlorobutane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68
Melting Pt: -132
Flash Pt: -10
MDL Number: MFCD00000871
CAS Number: 78-86-4
UN: 1127
ADR: 3 II
Merck Index: 13,01560

Order Now

Specification Test Results

Solubility 1.0 g/l (25 °C)
Melting Point -132 °C
Molar Mass 92.56 g/mol
Boiling Point 65 °C (1013 hPa)
Vapor Pressure 160 hPa (20 °C)
Flash Point -10 °C
Refractive Index 1.395 (20 °C, 589 nm)
Explosion Limit 2.0 - 8.8 %(V)
Density 0.87 g/cm3 (20 °C)
pH 7 (H2O, 20 °C) Aqueous solution
Ignition Point 460 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR