(±)-Camphene mixture of isomers for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20254.0100 8.20254.1000
CA8.20254.0100CS 357 CAD
CA8.20254.0100 CA8.20254.1000
(±)-Camphene mixture of isomers for synthesis, Sigma-Aldrich®
(±)-Camphene
Formula: C₁₀H₁₆
Boiling Pt: 158…159 °C (1013 hPa)
Melting Pt: 48…52 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00066603
CAS Number: 79-92-5
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Assay isomers (GC, area%) ≥ 90.0 % (a/a)
Assay (GC, Tricyclene) ≤ 10.0 % (a/a)
Identity (IR) passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
The product consists as a mixture of isomers >80,0 % camphene and >17,0 % fenchene.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR