2-Phenylaniline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20155.0010 8.20155.0050
CA8.20155.0010EA 79 CAD
CA8.20155.0010 CA8.20155.0050
2-Phenylaniline for synthesis, Sigma-Aldrich®
2-Phenylaniline
Formula: C₁₂H₁₁N
MDL Number: MFCD00007701
CAS Number: 90-41-5

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 46 - 48 °C
Molar Mass 169.23 g/mol
Boiling Point 298 - 300 °C
Flash Point 153 °C
Ignition Point 461 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR