Ethanolamine hydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Aminoethanol hydrochloride, 2-Hydroxyethylammonium Chloride, Monoethanolamine Hydrochloride, 2-Aminoethan-1-ol hydrochloride

M-Clarity™ quality level = MQ200

8.20063.0100 8.20063.0500
CA8.20063.0100EA 112 CAD
CA8.20063.0100 CA8.20063.0500
Ethanolamine hydrochloride for synthesis, Sigma-Aldrich®
Ethanolamine hydrochloride
Formula: C₂H₈ClNO
MW: 97.54 g/mol
MDL Number: MFCD00012892
CAS Number: 2002-24-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 80 - 84 °C
Molar Mass 97.54 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR