Silver tetrafluoroborate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18881.0005 8.18881.0025
CA8.18881.0005EA 218 CAD
CA8.18881.0005 CA8.18881.0025
Silver tetrafluoroborate for synthesis, Sigma-Aldrich®
Silver tetrafluoroborate
Formula: AgBF₄
MDL Number: MFCD00003408
CAS Number: 14104-20-2
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 70 - 73 °C
Molar Mass 194.67 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR