Ethyl palmitate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18775.0005 8.18775.0250
CA8.18775.0005EA 41 CAD
CA8.18775.0005 CA8.18775.0250
Ethyl palmitate for synthesis, Sigma-Aldrich®
Ethyl palmitate
Formula: C₁₈H₃₆O₂
MDL Number: MFCD00008996
CAS Number: 628-97-7

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 23 - 26 °C
Molar Mass 284.48 g/mol
Boiling Point 192 - 194 °C (13 hPa)
Flash Point 164 °C
Refractive Index 1.438 (25 °C, 589 nm)
Density 0.86 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR