1-Hexadecanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18704.0100 8.18704.1000
CA8.18704.0100CS 573 CAD
CA8.18704.0100 CA8.18704.1000
1-Hexadecanol for synthesis, Sigma-Aldrich®
1-Hexadecanol
Formula: H₃C(CH₂)₁₄CH₂OH
MDL Number: MFCD00004760
CAS Number: 36653-82-4

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (25 °C)
Melting Point 46 - 52 °C
Molar Mass 242.44 g/mol
Bulk Density 410 - 450 kg/m3
Boiling Point 305 - 330 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 155 °C DIN 51758
Density 0.82 g/cm3 (50 °C)
pH (H2O) not applicable
Ignition Point 235 °C DIN 51794

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR