Sodium-tert-butanolate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate

M-Clarity™ quality level = MQ200

8.18374.0100 8.18374.1000 8.18374.5000
CA8.18374.0100EA 88 CAD
CA8.18374.0100 CA8.18374.1000 CA8.18374.5000
Sodium-tert-butanolate for synthesis, Sigma-Aldrich®
Sodium tert-butanolate
Formula: C₄H₉NaO
MW: 96.1 g/mol
MDL Number: MFCD00040575
CAS Number: 865-48-5
UN: 3206
ADR: 4.2 (8),II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 263 °C
Molar Mass 96.1 g/mol
Bulk Density 400 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR