Semicarbazide hydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18369.0100 8.18369.0500
CA8.18369.0100CS 1172 CAD
CA8.18369.0100 CA8.18369.0500
Semicarbazide hydrochloride for synthesis, Sigma-Aldrich®
Semicarbazide hydrochloride
Formula: CH₆ClN₃O
MDL Number: MFCD00013009
CAS Number: 563-41-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 175 - 177 °C (decomposition)
Molar Mass 111.53 g/mol
Bulk Density 740 kg/m3
pH 1.5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR