3-Hydroxypropiononitrile for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Ethylene Cyanohydrin, 2-Cyanoethanol, 3-Hydroxypropionitrile, Hydracrylonitrile, β-Hydroxypropiononitrile
M-Clarity™ quality level = MQ200
8.18084.0100
8.18084.0500
CA8.18084.0100CS
760
CAD
CA8.18084.0100
CA8.18084.0500
3-Hydroxypropiononitrile for synthesis, Sigma-Aldrich®
3-Hydroxypropiononitrile
Formula:
C₃H₅NO MW: 71.08 g/mol Boiling Pt: 228 Melting Pt: -46 |
MDL Number:
MFCD00002826 CAS Number: 109-78-4 Merck Index: 13,03828 |
Specification Test Results
Solubility | 1000 g/l (20 °C) |
Melting Point | -46 °C |
Molar Mass | 71.08 g/mol |
Boiling Point | 228 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 128 °C DIN 51758 |
Refractive Index | 1.4241 (25 °C, 589 nm) |
Density | 1.05 g/cm3 (20 °C) |
pH | 3.0 - 4.5 (H2O, 20 °C) (undiluted) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...