Benzyl butyrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.16125.0100 8.16125.0500
CA8.16125.0100CS 992 CAD
CA8.16125.0100 CA8.16125.0500
Benzyl butyrate for synthesis, Sigma-Aldrich®
Benzyl butyrate
Formula: C₁₁H₁₄O₂
MDL Number: MFCD00027133
CAS Number: 103-37-7

Order Now

Specification Test Results

Solubility (20 °C) low
Molar Mass 178.23 g/mol
Boiling Point 235 - 242 °C (1013 hPa)
Vapor Pressure 11.97 hPa (109 °C)
Flash Point 107 °C
Density 1.01 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR