L(+)-Cysteine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: H-Cys-OH, (R)-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

M-Clarity™ quality level = MQ200

8.16014.0025 8.16014.0100
CA8.16014.0025EA 54 CAD
CA8.16014.0025 CA8.16014.0100
L(+)-Cysteine for synthesis, Sigma-Aldrich®
L(+)-Cysteine
Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
MDL Number: MFCD00064306
CAS Number: 52-90-4

Order Now

Specification Test Results

Solubility 160 g/l (20 °C)
Melting Point 220 - 228 °C
Molar Mass 121.16 g/mol
Bulk Density 300 kg/m3
pH 4.5 - 5.5 (100 g/l, H2O, 20 °C)
Ignition Point 420 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR