L(+)-Valine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.16006.0025 8.16006.0100
CA8.16006.0025EA 109 CAD
CA8.16006.0025 CA8.16006.0100
L(+)-Valine for synthesis, Sigma-Aldrich®
L(+)-Valine
Formula: C₅H₁₁NO₂
MDL Number: MFCD00064220
CAS Number: 72-18-4

Order Now

Specification Test Results

Solubility 85 g/l (20 °C)
Melting Point 315 °C
Molar Mass 117.15 g/mol
Bulk Density 400 kg/m3
Density 1.23 g/cm3 (20 °C)
pH 5.5 - 6.5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR