Methyl enantate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.14926.0100
8.14926.0500
CA8.14926.0100CS
1244
CAD
CA8.14926.0100
CA8.14926.0500
Methyl enantate for synthesis, Sigma-Aldrich®
Methyl enantate
Formula:
C₈H₁₆O₂ |
MDL Number:
MFCD00009537 CAS Number: 106-73-0 UN: 3272 ADR: 3,III |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | -56 °C |
Molar Mass | 144.21 g/mol |
Boiling Point | 173 °C (1013 hPa) |
Flash Point | 57 °C |
Density | 0.880 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...