Isopentyl propionate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14845.0025 8.14845.0100
CA8.14845.0025EA 48 CAD
CA8.14845.0025 CA8.14845.0100
Isopentyl propionate for synthesis, Sigma-Aldrich®
Isopentyl propionate
Formula: C₈H₁₆O₂
MDL Number: MFCD00048611
CAS Number: 105-68-0
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Melting Point -73 °C
Molar Mass 144.21 g/mol
Boiling Point 160 °C
Flash Point 46 °C
Explosion Limit 1 %(V)
Density 0.869 g/cm3 (20 °C)
Ignition Point 370 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR