p,p,p-Tritolylphosphate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14811.0250 8.14811.1000
CA8.14811.0250CS 1178 CAD
CA8.14811.0250 CA8.14811.1000
p,p,p-Tritolylphosphate for synthesis, Sigma-Aldrich®
p,p,p-Tritolyl phosphate
Formula: C₂₁H₂₁O₄P
CAS Number: 78-32-0
UN: 2574
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 368.37 g/mol
Boiling Point 300 °C
Vapor Pressure 0.044 hPa (150 °C)
Flash Point 220 °C
Density 1.17 g/cm3 (20 °C)
Viscosity kinematic 70 mm2/s (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR