(R)-(+)-1,1'-Bi-2-naphthol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: (R)-[1,1'-Binaphthalene]-2,2'-diol, (R)-BINOL, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol

M-Clarity™ quality level = MQ200

8.14774.0001 8.14774.0010
CA8.14774.0001EA 56 CAD
CA8.14774.0001 CA8.14774.0010
(R)-(+)-1,1'-Bi-2-naphthol for synthesis, Sigma-Aldrich®
(R)-(+)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
MDL Number: MFCD00004068
CAS Number: 18531-94-7
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 204 - 208 °C
Molar Mass 286.33 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR