Diisodecyl phthalate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14732.0100 8.14732.1000
CA8.14732.0100CS 2109 CAD
CA8.14732.0100 CA8.14732.1000
Diisodecyl phthalate for synthesis, Sigma-Aldrich®
Diisodecyl phthalate
Formula: C₂₈H₄₆O₄
MDL Number: MFCD00048389
CAS Number: 26761-40-0

Order Now

Specification Test Results

Solubility 0.00028 g/l (24 °C)
Melting Point -50 °C
Molar Mass 446.66 g/mol
Boiling Point 250 - 267 °C (7 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 240 °C
Density 0.97 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 380 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR