Methanesulfonic acid ≥70% in water for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14637.0250 8.14637.1000
CA8.14637.0250CS 732 CAD
CA8.14637.0250 CA8.14637.1000
Methanesulfonic acid ≥70% in water for synthesis, Sigma-Aldrich®
Methanesulphonic acid
Formula: CH₃SO₃H
MDL Number: MFCD00007518
CAS Number: 75-75-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -60 °C
Density 1.35 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR