1,2,4-Trimethylbenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14505.0100 8.14505.0500
CA8.14505.0100CS 597 CAD
CA8.14505.0100 CA8.14505.0500
1,2,4-Trimethylbenzene for synthesis, Sigma-Aldrich®
1,2,4-Trimethylbenzene
Formula: C₉H₁₂
MDL Number: MFCD00008527
CAS Number: 95-63-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Solubility 0.057 g/l (20 °C)
Melting Point -44 °C
Molar Mass 120.19 g/mol
Boiling Point 168 °C (1013 hPa)
Vapor Pressure 2.8 hPa (25 °C)
Flash Point 48 °C
Refractive Index 1.5047 (20 °C, 589 nm)
Explosion Limit 0.8 - 7 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 485 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR